missing translation for 'onlineSavingsMsg'
Learn More

SLC5A3 Antibody, Novus Biologicals™

Product Code. 18389351
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389351 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18389351 Supplier Bio-Techne Supplier No. NBP314475

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC5A3 Polyclonal antibody specifically detects SLC5A3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC5A3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias human solute carrier family 5, member 3, Sodium, human solute carrier family 5, member 3, Sodium/myo-inositol cotransporter, myo-inositol cotransporter10, Na(+)/myo-inositol cotransporter, SMIT, SMIT2, sodium/myo-inositol cotransporter, sodium/myo-inositol cotransporter 1, solute carrier family 5 (inositol transporter), member 3, solute carrier family 5 (inositol transporters), member 3, solute carrier family 5 (sodium/myo-inositol cotransporter), member 3, Solute carrier family 5 member 3
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from internal region of human SLC5A3.
Purification Method Immunogen affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 6526
Target Species Human
Content And Storage Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Product Type Antibody
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.