missing translation for 'onlineSavingsMsg'
Learn More

SLC5A3 Antibody (3A6), Novus Biologicals™

Product Code. 18391849 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18391849 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18391849 Supplier Novus Biologicals Supplier No. H00006526M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SLC5A3 Monoclonal antibody specifically detects SLC5A3 in Human, Mouse samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC5A3
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3A6
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_008864
Gene Alias human solute carrier family 5, member 3, Sodium, human solute carrier family 5, member 3, Sodium/myo-inositol cotransporter, myo-inositol cotransporter10, Na(+)/myo-inositol cotransporter, SMIT, SMIT2, sodium/myo-inositol cotransporter, sodium/myo-inositol cotransporter 1, solute carrier family 5 (inositol transporter), member 3, solute carrier family 5 (inositol transporters), member 3, solute carrier family 5 (sodium/myo-inositol cotransporter), member 3, Solute carrier family 5 member 3
Host Species Mouse
Immunogen SLC5A3 (NP_008864, 533 a.a. ~ 641 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. TPPPTKEQIRTTTFWSKKNLVVKENCSPKEEPYKMQEKSILRCSENNETINHIIPNGKSEDSIKGLQPEDVNLLVTCREEGNPVASLGHSEAETPVDAYSNGQAALMGE
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 6526
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.