missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC5A12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110389
This item is not returnable.
View return policy
Description
Immunogen sequence: LIQGSTHAGG FHNVLEQSTN GSRLHIFDFD VDPLRRHTFW TITVGGTFTW LGIYGVNQST IQRCISCKTE KHAKLALYFN L.
Normal blood lactate is maintained at about 1.5 mM, and little filtered lactate is excreted in urine. Reabsorption of lactate is mediated by the low-affinity Na(+)-coupled lactate transporter SLC5A12 in the initial part of the proximal tubule and by the high-affinity Na(+)-coupled lactate transporter SLC5A8 (MIM 608044) in the distal proximal tubule (Gopal et al., 2007 [PubMed 17692818]).
Specifications
| SLC5A12 | |
| Polyclonal | |
| Unconjugated | |
| SLC5A12 | |
| electroneutral sodium monocarboxylate cotransporter; Low-affinity sodium-lactate cotransporter; SLC5A12; SMCT2; Sodium-coupled monocarboxylate transporter 2; sodium-iodide related cotransporter; solute carrier family 5 (sodium/glucose cotransporter), member 12; solute carrier family 5 (sodium/monocarboxylate cotransporter), member 12; solute carrier family 5 member 12 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 159963 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.97 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q1EHB4 | |
| SLC5A12 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction