missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC5A12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-49322-25ul
This item is not returnable.
View return policy
Description
SLC5A12 Polyclonal antibody specifically detects SLC5A12 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| SLC5A12 | |
| Polyclonal | |
| Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/ Immunofluorescence -Reported in scientific literature (PMID: 31708446), Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Electroneutral sodium monocarboxylate cotransporter, Low-affinity sodium-lactate cotransporter, MGC52019, SMCT2DKFZp564G223, sodium-coupled monocarboxylate transporter 2, sodium-iodide related cotransporter, solute carrier family 5 (sodium/glucose cotransporter), member 12, Solute carrier family 5 member 12 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: QGSTHAGGFHNVLEQSTNGSRLHIFDFDVDPLRRH | |
| 25 μL | |
| Cancer, Endocrinology, Neuroscience, Signal Transduction | |
| 159963 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction