missing translation for 'onlineSavingsMsg'
Learn More

SLC5A11 Antibody, Novus Biologicals™

Product Code. 18327672
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18327672 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18327672 Supplier Bio-Techne Supplier No. NBP314474

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC5A11 Polyclonal antibody specifically detects SLC5A11 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC5A11
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias KST1RKST1, Na(+)/myo-inositol cotransporter 2, na+/myo-inositol cotransporter 2, putative sodium-coupled cotransporter RKST1, SGLT6, SLGTX, SMIT2homolog of rabbit KST1, Sodium/glucose cotransporter KST1, sodium/myo-inositol cotransporter 2, Sodium-dependent glucose cotransporter, solute carrier family 5 (sodium/glucose cotransporter), member 11, Solute carrier family 5 member 11
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from internal region of human SLC5A11.
Purification Method Immunogen affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 115584
Target Species Human
Content And Storage Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Product Type Antibody
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.