missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC52A3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25626-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| SLC52A3 | |
| Polyclonal | |
| Unconjugated | |
| SLC52A3 | |
| C20orf54, hRFT2, RFT2, RFVT3, Riboflavin transporter 2, SLC52A3, Solute carrier family 52, riboflavin transporter, member 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 113278 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9NQ40 | |
| SLC52A3 | |
| SLC52A3 Fusion Protein Ag22315 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction