missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC52A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564086
This item is not returnable.
View return policy
Description
Immunogen sequence: ELGSGLQVGA PGAEEEVEES SPL Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 56%.
This gene encodes a membrane protein which belongs to the riboflavin transporter family. In humans, riboflavin must be obtained by intestinal absorption because it cannot be synthesized by the body. The water-soluble vitamin riboflavin is processed to the coenzymes flavin mononucleotide (FMN) and flavin adenine dinucleotide (FAD) which then act as intermediaries in many cellular metabolic reactions. Paralogous members of the riboflavin transporter gene family are located on chromosomes 17 and 20. Unlike other members of this family, this gene has higher expression in brain tissue than small intestine. Alternative splicing of this gene results in multiple transcript variants encoding the same protein. Mutations in this gene have been associated with Brown-Vialetto-Van Laere syndrome 2 - an autosomal recessive progressive neurologic disorder characterized by deafness, bulbar dysfunction, and axial and limb hypotonia. [provided by RefSeq, Jul 2012].
Specifications
| SLC52A2 | |
| Polyclonal | |
| Unconjugated | |
| SLC52A2 | |
| 2010003P03Rik; BVVLS2; D15Ertd747e; G protein-coupled receptor 172A; G protein-coupled receptor 172B; GPCR41; GPCR42; Gpr172a; Gpr172b; hRFT3; mRFT1; PAR1; PAR2; PERV-A receptor 1; PERV-A receptor 2 homolog; PEVR1; Porcine endogenous retrovirus A receptor 1; porcine endogenous retrovirus A receptor 2; Porcine endogenous retrovirus A receptor 2 homolog; Protein GPR172A; Protein GPR172B; putative G-protein coupled receptor GPCR41; Rft1; RFT3; RFVT2; RGD1560410; riboflavin transporter 1; Riboflavin transporter 3; rRFT1; Slc52a2; solute carrier family 52 (riboflavin transporter), member 2; solute carrier family 52 member 2; solute carrier family 52, riboflavin transporter, member 2; solute carrier protein 52, member 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79581 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HAB3 | |
| SLC52A2 | |
| Recombinant protein corresponding to Human SLC52A2. Recombinant protein control fragment (Product #RP-104034). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction