missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC50A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570596
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 86%; Dog: 79%; Guinea Pig: 86%; Horse: 91%; Human: 100%; Mouse: 79%; Pig: 93%; Rabbit: 86%; Rat: 86%.
SLC50A1 gene ontology annotations related to this gene include carbohydrate transport.
Specifications
| SLC50A1 | |
| Polyclonal | |
| Unconjugated | |
| SLC50A1 | |
| HsSWEET1; LOW QUALITY PROTEIN: sugar transporter SWEET1; MmSWEET1; probable sugar transporter RAG1AP1; RAG1-activating protein 1; RAG1AP1; recombination activating gene 1 activating protein 1; recombination activating gene 1 gene activation; Rga; RGD1308478; RP11-540D14.5; RZPDo834D038D; SCP; SLC50A1; slv; solute carrier family 50 (sugar efflux transporter), member 1; solute carrier family 50 (sugar transporter), member 1; solute carrier family 50 member 1; Stromal cell protein; sugar transporter SWEET1; SWEET1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 55974 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BRV3 | |
| SLC50A1 | |
| synthetic peptide directed towards the middle region of human SLC50A1 (aa 139-188). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction