missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC4A8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101904
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SLC4A8/10.
Mediates electroneutral sodium- and carbonate-dependent choride-HCO3(-) exchange with a Na(+):HCO3(-) stoichiometry of 2:1. Plays a major role in pH regulation in neurons. May be involved in cell pH regulation by transporting HCO3(-) from blood to cell. Enhanced expression in severe acid stress could be important for cell survival by mediating the influx of HCO3(-) into the cells. Also mediates lithium-dependent HCO3(-) cotransport. May be regulated by osmolarity.
Specifications
| SLC4A8 | |
| Polyclonal | |
| Unconjugated | |
| Slc4a8 | |
| AW493845; C230026C11; Electroneutral Na(+)-driven Cl-HCO3 exchanger; electroneutral Na+-driven Cl-HCO3 exchanger; Electroneutral sodium bicarbonate exchanger 1; KIAA0739; k-NBC3; kNBC-3; Na-driven Cl-HCO3 exchanger NDCBE1-A; Na-driven Cl-HCO3 exchanger NDCBE1-B; Na-driven Cl-HCO3 exchanger NDCBE1-C; NBC; NBC3; NDCBE; NDCBE1; RGD:735164}; Slc4a8; slc4a8 {ECO:0000312; sodium bicarbonate cotransporter isoform 3 kNBC-3; solute carrier family 4 (anion exchanger), member 8; solute carrier family 4 member 8; solute carrier family 4, sodium bicarbonate cotransporter, member 8 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 315311, 59033, 9498 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q2Y0W8, Q6RVG2, Q8JZR6 | |
| Slc4a8 | |
| A synthesized peptide derived from human SLC4A8(Accession Q2Y0W8), corresponding to amino acid residues F1040-S1090. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction