missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC4A8, Mouse anti-Human, Clone: 6E11, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00009498-M04.100ug
This item is not returnable.
View return policy
Description
Sequence: IEEISDLILDQQELSSDLNDSMRVKVREALLKKHHHQNEKKRNNLIPIVRSFAEVGKKQSDPHLMDKHGQTVSPQSVPTTNLEVKNGVNCEHSPVDLSKVSpecifications
| SLC4A8 | |
| Monoclonal | |
| Unconjugated | |
| solute carrier family 4, sodium bicarbonate cotransporter, member 8 | |
| DKFZp761B2318/FLJ46462/NBC3 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 6E11 | |
| In 1x PBS, pH 7.4 | |
| NM_004858 | |
| SLC4A8 | |
| SLC4A8 (NP_004849, 187 a.a. ∼ 286 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 9498 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG3 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction