missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC4A4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £443.00
Specifications
| Antigen | SLC4A4 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18410462
|
Novus Biologicals
NBP2-32021-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18143255
|
Novus Biologicals
NBP2-32021 |
0.1 mL |
£443.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC4A4 Polyclonal specifically detects SLC4A4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SLC4A4 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| electrogenic sodium bicarbonate cotransporter 1, hhNMC, HNBC1, KNBC, kNBC1, Na(+)/HCO3(-) cotransporter, NBC, NBC1DKFZp781H1314, NBC2, NBCE1, pNBC, SLC4A5, Sodium bicarbonate cotransporter, sodium bicarbonate cotransporter 1 (sodium bicarbonate cotransporter, kidney;sodium bicarbonate cotransporter, pancreas), Solute carrier family 4 member 4, solute carrier family 4, sodium bicarbonate cotransporter, member 4, solute carrier family 4, sodium bicarbonate cotransporter, member 4, brain type, solute carrier family 4, sodium bicarbonate cotransporter, member 5 | |
| SLC4A4 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q9Y6R1 | |
| 8671 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: DYYPINSNFKVGYNTLFSCTCVPPDPANISISNDTTLAPEYLPTMSSTDMYHNTTFDWAFLSKKECSKYGGNLVGNN | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title