missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC4A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568092
This item is not returnable.
View return policy
Description
Predicted to react with Mouse samples.
Plasma membrane anion exchange protein of wide distribution.
Specifications
| SLC4A2 | |
| Polyclonal | |
| Unconjugated | |
| SLC4A2 | |
| AE 2; AE2; Aep2; anion exchange protein 2; anion exchanger 2; anion exchanger 2 type a; anion exchanger 2 type b1; anion exchanger 2 type b2; B3RP; B3rp2; B3RP-2; Band 3-related protein; band 3-related protein 2; BND3L; EPB3L1; Erythrocyte band 3; erythrocyte membrane protein band 3-like 1; HKB3; MPB3L; NBND3; non-erythroid band 3-like protein; Slc4a2; solute carrier family 4 (anion exchanger), member 2; solute carrier family 4 member 2; Solute carrier family 4, member 2, anion exchange protein 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20535, 24780, 6522 | |
| -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P04920, P13808, P23347 | |
| SLC4A2 | |
| Synthesized peptide derived from human SLC4A2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction