Learn More
Invitrogen™ SLC4A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534206
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Orangutan, Gibbon, Galago, Marmoset, Mouse, Rat, Hamster, Panda, Dog, Horse, Rabbit, Pig (100%); Monkey, Bovine, Guinea pig (94%); Elephant (89%); Bat, Opossum (83%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
Plasma membrane anion exchange protein of wide distribution.
Specifications
| SLC4A2 | |
| Polyclonal | |
| Unconjugated | |
| SLC4A2 | |
| AE 2; AE2; Aep2; anion exchange protein 2; anion exchanger 2; anion exchanger 2 type a; anion exchanger 2 type b1; anion exchanger 2 type b2; B3RP; B3rp2; B3RP-2; Band 3-related protein; band 3-related protein 2; BND3L; EPB3L1; Erythrocyte band 3; erythrocyte membrane protein band 3-like 1; HKB3; MPB3L; NBND3; non-erythroid band 3-like protein; Slc4a2; solute carrier family 4 (anion exchanger), member 2; solute carrier family 4 member 2; Solute carrier family 4, member 2, anion exchange protein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100009159, 100762427, 20535, 24780, 397203, 475538, 6522, 791243 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with 0.1% sodium azide | |
| P04920, P13808, P23347, P48746, Q6SJP2 | |
| SLC4A2 | |
| Synthetic 18 amino acid peptide from N-Terminus of human SLC4A2. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Rabbit, Hamster, Pig, Horse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.