missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC4A11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101889
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SLC4A11.
This gene encodes a voltage-regulated, electrogenic sodium-coupled borate cotransporter that is essential for borate homeostasis, cell growth and cell proliferation. Mutations in this gene have been associated with a number of endothelial corneal dystrophies including recessive corneal endothelial dystrophy 2, corneal dystrophy and perceptive deafness, and Fuchs endothelial corneal dystrophy. Multiple transcript variants encoding different isoforms have been described.
Specifications
| SLC4A11 | |
| Polyclonal | |
| Unconjugated | |
| SLC4A11 | |
| AI503023; bicarbonate transporter related protein 1; bicarbonate transporter-related protein 1; BTR1; CDPD; CDPD1; CHED; CHED2; dJ794I6.2; MGC126418; MGC126419; NABC1; OTTHUMP00000030097; Slc4a11; Sodium bicarbonate transporter-like protein 11; Sodium borate cotransporter 1; sodium-coupled borate cotransporter 1; solute carrier family 4 member 11; solute carrier family 4, sodium bicarbonate transporter-like, member 11; solute carrier family 4, sodium borate transporter, member 11 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 269356, 83959 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A2AJN7, Q8NBS3 | |
| SLC4A11 | |
| A synthesized peptide derived from human SLC4A11(Accession Q8NBS3), corresponding to amino acid residues E293-G343. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion