Learn More
Invitrogen™ SLC4A11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590811
Description
Positive Samples: 293T, Mouse kidney; Cellular Location: Cell membrane, Membrane, Multi-pass membrane protein Immunogen sequence: MAAATRRVFH LQPCENSPTM SQNGYFEDSS YYKCDTDDTF EAREEILGDE AFDTANSSIV SGESIRFFVN VNLEMQATNT ENEATSGGCV LLHTSRKYLK LKNFKEEIRA HRDLDGFLAQ ASIVLNETAT SLDNVLRTML RRFARDPDNN EPNCNLDLLM AMLFTDAGAP MRGKVHLLSD.
This gene encodes a voltage-regulated, electrogenic sodium-coupled borate cotransporter that is essential for borate homeostasis, cell growth and cell proliferation. Mutations in this gene have been associated with a number of endothelial corneal dystrophies including recessive corneal endothelial dystrophy 2, corneal dystrophy and perceptive deafness, and Fuchs endothelial corneal dystrophy. Multiple transcript variants encoding different isoforms have been described.
Specifications
| SLC4A11 | |
| Polyclonal | |
| Unconjugated | |
| SLC4A11 | |
| AI503023; bicarbonate transporter related protein 1; bicarbonate transporter-related protein 1; BTR1; CDPD; CDPD1; CHED; CHED2; dJ794I6.2; MGC126418; MGC126419; NABC1; OTTHUMP00000030097; Slc4a11; Sodium bicarbonate transporter-like protein 11; Sodium borate cotransporter 1; sodium-coupled borate cotransporter 1; solute carrier family 4 member 11; solute carrier family 4, sodium bicarbonate transporter-like, member 11; solute carrier family 4, sodium borate transporter, member 11 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 269356, 83959 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.6 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| A2AJN7, Q8NBS3 | |
| SLC4A11 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human SLC4A11 (NP_0011675601). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.