Learn More
SLC4A10 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27197-1-AP-20UL
Description
The SLC4A10 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SLC4A10. This antibody recognizes human, mouse antigen. The SLC4A10 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| SLC4A10 | |
| 550 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| NBCn2, NCBE, SLC4A10 | |
| 20 μL | |
| 57282 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |