missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC4A10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98632-50ul
This item is not returnable.
View return policy
Description
SLC4A10 Polyclonal antibody specifically detects SLC4A10 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SLC4A10 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| sodium bicarbonate cotransporter-like, member 10, sodium bicarbonate transporter-like, member 10, sodium-driven chloride bicarbonate exchanger, Solute carrier family 4 member 10, solute carrier family 4, sodium bicarbonate transporter, member 10 | |
| Produced in rabbits immunized with E. coli-derived Human SLC4A10 fragment. | |
| 50 μg | |
| Neuroscience | |
| 57282 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction