missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC47A2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA537990
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
S47A2 belongs to a family of transporters involved in excretion of toxic electrolytes, both endogenous and exogenous, through urine and bile. This transporter family shares homology with the bacterial MATE (multidrug and toxin extrusion) protein family responsible for drug resistance. This gene is one of two members of the MATE transporter family located near each other on chromosome 17.
Specifications
| SLC47A2 | |
| Polyclonal | |
| Unconjugated | |
| Slc47a2 | |
| 4933429E10Rik; FLJ31196; H(+)/organic cation antiporter kidney-specific; hMATE-2; kidney-specific H(+)/organic cation antiporter; Mate2; MATE-2; MATE2-B; MATE2K; MATE2-K; mMATE-2; multidrug and toxin extrusion protein 2; SLC47A2; solute carrier family 47 (multidrug and toxin extrusion), member 2; solute carrier family 47 member 2; solute carrier family 47, member 2 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 146802 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q86VL8 | |
| Slc47a2 | |
| Peptide with sequence C-QRAESTATRPGPEK, from the internal region near the C-terminus of SLC47A2 (aa 511-525). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction