missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC47A1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA519085
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
This gene is located within the Smith-Magenis syndrome region on chromosome 17. It encodes a protein of unknown function.
Specifications
| SLC47A1 | |
| Polyclonal | |
| Unconjugated | |
| Slc47a1 | |
| 1300013J15Rik; AI036982; FLJ10847; H+/organic cation antiporter variant 1; H+/organic cation antiporter variant 2; hMATE-1; MATE1; MATE-1; MGC64822; mMATE1; mMATE-1; multidrug and toxin extrusion 1; multidrug and toxin extrusion protein 1; RGD1311123; rMATE-1; SLC47A1; solute carrier family 47 (multidrug and toxin extrusion), member 1; solute carrier family 47 member 1; solute carrier family 47, member 1 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 55244 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q96FL8 | |
| Slc47a1 | |
| Synthetic peptide sequence (PEHPQDGAKLSRKQ) corresponding to the internal amino acids of SLC47A1 (aa 528-542). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction