missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC46A1 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142358
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.
Has been shown to act both as an intestinal proton-coupled high-affinity folate transporter and as an intestinal heme transporter which mediates heme uptake from the gut lumen into duodenal epithelial cells. The iron is then released from heme and may be transported into the bloodstream. Dietary heme iron is an important nutritional source of iron. Shows a higher affinity for folate than heme.
Specifications
| SLC46A1 | |
| Polyclonal | |
| Unconjugated | |
| SLC46A1 | |
| 1110002C08Rik; D11Ertd18e; G21; Hcp1; Heme carrier protein 1; Pcft; PCFT/HCP1; proton-coupled folate transporter; retinal pigment epithelilum transporter; retinal pigment epithelium transporter; RGD1309472; Slc46a1; solute carrier family 46 (folate transporter), member 1; solute carrier family 46 member 1; solute carrier family 46, member 1; TRPE; unnamed protein product | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 113235 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q96NT5 | |
| SLC46A1 | |
| Peptide with sequence YNGTRQRGGCSNRS. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction