missing translation for 'onlineSavingsMsg'
Learn More

SLC45A4 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Thermo Scientific PA554711

Additional Details : Weight : 0.02000kg

Product Code. 15742641

  • £428.00 / 100µL
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Immunogen sequence: NVSEEAKEEQ KGLSSPLAGE GRAGGNSEKP TVLKLTRKEG LQGPVETERL QVLTSVRSRH IGWCR Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 54%.

SLC45A4 Polyclonal antibody specifically detects SLC45A4 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

SLC45A4
Polyclonal
Unconjugated
SLC45A4
KIAA1126; SLC45A4; Solute carrier family 45 member 4; solute carrier family 45, member 4
Rabbit
Antigen affinity chromatography
RUO
57210
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q5BKX6
SLC45A4
Recombinant protein corresponding to Human SLC45A4
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only