missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC45A4 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5148321

Product Code. 30019550

  • £313.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SLC45A4 Polyclonal Antibody for IHC (P)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLC45A4
Polyclonal
Unconjugated
SLC45A4
KIAA1126; SLC45A4; Solute carrier family 45 member 4; solute carrier family 45, member 4
Rabbit
Antigen affinity chromatography, Protein A
RUO
57210
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
100 μg/mL
PBS with 0.03% ProClin 300; pH 7
Q5BKX6
SLC45A4
E. coli-derived Human SLC45A4 fragment.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.