missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SLC45A3/Prostein Monoclonal specifically detects SLC45A3/Prostein in Human samples. It is validated for Immunohistochemistry.
Specifications
Specifications
| Antigen | SLC45A3/Prostein |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Clone | RM426 |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:100-1:400 |
| Formulation | PBS, 50% Glycerol, 1% BSA |
| Gene Alias | IPCA-2, IPCA-6, IPCA-8, PCANAP6IPCA6, PCANAP8, prostate cancer associated protein 2, prostate cancer associated protein 6, prostate cancer associated protein 8, prostate cancer-associated gene 2, prostate cancer-associated gene 6, prostate cancer-associated gene 8, Prostate cancer-associated protein 6, Prostein, PRSTPCANAP2, solute carrier family 45 member 3, solute carrier family 45, member 3 |
| Host Species | Rabbit |
| Immunogen | A peptide corresponding to the C-terminus of human SLC45A3/Prostein |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?