missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC45A3/Prostein Rabbit anti-Human, Clone: RM426, Novus Biologicals™
Rabbit Monoclonal Antibody
£155.00 - £416.00
Specifications
| Antigen | SLC45A3/Prostein |
|---|---|
| Clone | RM426 |
| Dilution | Immunohistochemistry 1:100-1:400 |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18372316
|
Novus Biologicals
NBP3-09178-25UL |
25 μg |
£155.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18396623
|
Novus Biologicals
NBP3-09178-100UL |
100 μg |
£416.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC45A3/Prostein Monoclonal specifically detects SLC45A3/Prostein in Human samples. It is validated for Immunohistochemistry.Specifications
| SLC45A3/Prostein | |
| Immunohistochemistry 1:100-1:400 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, 50% Glycerol, 1% BSA | |
| 85414 | |
| IgG | |
| Protein A purified |
| RM426 | |
| Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| IPCA-2, IPCA-6, IPCA-8, PCANAP6IPCA6, PCANAP8, prostate cancer associated protein 2, prostate cancer associated protein 6, prostate cancer associated protein 8, prostate cancer-associated gene 2, prostate cancer-associated gene 6, prostate cancer-associated gene 8, Prostate cancer-associated protein 6, Prostein, PRSTPCANAP2, solute carrier family 45 member 3, solute carrier family 45, member 3 | |
| A peptide corresponding to the C-terminus of human SLC45A3/Prostein | |
| Primary | |
| Store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title