Learn More
Invitrogen™ SLC44A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143941
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: mouse lung tissue. IHC: mouse intestine tissue. Flow: HEPA1-6 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Isoform 1, but not isoform 3, exhibits some choline transporter activity.
Specifications
| SLC44A2 | |
| Polyclonal | |
| Unconjugated | |
| SLC44A2 | |
| 1110028E10Rik; Choline transporter-like protein 2; Ctl2; IESCA; inner ear supporting cell antigen; PP1292; PSEC0210; RGD1309680; SLC44A2; solute carrier family 44 (choline transporter), member 2; solute carrier family 44 member 2; solute carrier family 44, member 2; solute carrier family 44, member 2 tv2; testicular tissue protein Li 47 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 363024, 68682 | |
| -20°C | |
| Lyophilized |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| B4F795, Q8BY89 | |
| SLC44A2 | |
| E.coli-derived mouse SLC44A2 recombinant protein (Position: M1-S706). | |
| 100 μg | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.