missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC44A2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94586-0.1ml
This item is not returnable.
View return policy
Description
SLC44A2 Polyclonal antibody specifically detects SLC44A2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| SLC44A2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CTL2choline transporter-like protein 2, DKFZp666A071, FLJ44586, PP1292, Solute carrier family 44 member 2, solute carrier family 44, member 2 | |
| A synthetic peptide corresponding to a sequence within amino acids 150-250 of human SLC44A2 (NP_065161.3). AEVLQDGDCPAVLIPSKPLARRCFPAIHAYKGVLMVGNETTYEDGHGSRKNITDLVEGAKKANGVLEARQLAMRIFEDYTVSWYWIIIGLVIAMAMSLLFI | |
| 0.1 mL | |
| Signal Transduction | |
| 57153 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction