missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC43A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA523571
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine based on sequence homology.
Uniporter that mediates the transport of the stereospecific L-phenylalanine, L-methionine and L-branched-chain amino acids, between the extracellular space and the cytoplasm and may control the transepithelial (re)absorption of neutral amino acid in kidney and small intestine. The transport activity is mediated through facilitated diffusion and is sodium ions-, chloride ions- and pH-independent.
Specifications
| SLC43A2 | |
| Polyclonal | |
| Unconjugated | |
| SLC43A2 | |
| 7630402D21Rik; BC042513; large neutral amino acids transporter small subunit 4; Lat4; L-type amino acid transporter 4; L-type amino L-acid transporter 4; PP7664; Slc43a2; solute carrier family 43 (amino acid system L transporter), member 2; solute carrier family 43 member 2; solute carrier family 43, member 2 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 124935, 215113 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8CGA3, Q8N370 | |
| SLC43A2 | |
| KLH conjugated synthetic peptide between 236-265 amino acids from the central region of human SLC43A2. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction