missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC43A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554451
This item is not returnable.
View return policy
Description
Immunogen sequence: YRRQLERQLQ QRQEDDKLFL KINGSSNQEA FV Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 91%.
Uniporter that mediates the transport of the stereospecific L-phenylalanine, L-methionine and L-branched-chain amino acids, between the extracellular space and the cytoplasm and may control the transepithelial (re)absorption of neutral amino acid in kidney and small intestine. The transport activity is mediated through facilitated diffusion and is sodium ions-, chloride ions- and pH-independent.
Specifications
| SLC43A2 | |
| Polyclonal | |
| Unconjugated | |
| SLC43A2 | |
| 7630402D21Rik; BC042513; large neutral amino acids transporter small subunit 4; Lat4; L-type amino acid transporter 4; L-type amino L-acid transporter 4; PP7664; Slc43a2; solute carrier family 43 (amino acid system L transporter), member 2; solute carrier family 43 member 2; solute carrier family 43, member 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 124935 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N370 | |
| SLC43A2 | |
| Recombinant protein corresponding to Human SLC43A2. Recombinant protein control fragment (Product #RP-93382). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction