Learn More
Invitrogen™ SLC43A1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5101071
Product Code. 16607554
  • £450.00 / 100µL
Description
Antibody detects endogenous levels of total LAT3.
SLC43A1 belongs to the system L family of plasma membrane carrier proteins that transports large neutral amino acids. SLC43A1 belongs to the system L family of plasma membrane carrier proteins that transports large neutral amino acids.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SLC43A1
Polyclonal
Unconjugated
SLC43A1
2610016F07Rik; AA986141; large neutral amino acids transporter small subunit 3; LAT3; L-type amino acid transporter 3; PB39; POV1; prostate cancer overexpressed gene 1; prostate cancer overexpressed gene 1 protein; R00504; Slc43a1; solute carrier family 43 (amino acid system L transporter), member 1; solute carrier family 43 member 1; solute carrier family 43, member 1
Rabbit
Affinity chromatography
RUO
72401, 8501
-20°C
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O75387, Q8BSM7
SLC43A1
A synthesized peptide derived from human SLC43A1(Accession O75387), corresponding to amino acid residues V245-L295.
100 μL
Primary
Human, Mouse