missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC41A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59693
This item is not returnable.
View return policy
Description
SLC41A1 Polyclonal specifically detects SLC41A1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| SLC41A1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| MgtE, solute carrier family 41 member 1, solute carrier family 41, member 1 | |
| Rabbit | |
| 55 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunoprecipitation | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunoprecipitation | |
| Q8IVJ1 | |
| SLC41A1 | |
| Synthetic peptides corresponding to SLC41A1(solute carrier family 41, member 1) The peptide sequence was selected from the N terminal of SLC41A1. Peptide sequence TSEFLGPDGAGVEVVIESRANAKGVREEDALLENGSQSNESDDVSTDRGP. | |
| Affinity purified | |
| RUO | |
| 254428 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction