Learn More
SLC3A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16343-1-AP-150UL
 View more versions of this product
Product Code. 16875934
  • £349.00 / 150µL
Description
This gene encodes a type II membrane glycoprotein which is one of the components of the renal amino acid transporter which transports neutral and basic amino acids in the renal tubule and intestinal tract. Mutations and deletions in this gene are associated with cystinuria. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
Specifications
Specifications
SLC3A1
Polyclonal
Unconjugated
SLC3A1
ATR1, CSNU1, D2H, NBAT, RBAT, SLC3A1
Rabbit
Antigen Affinity Chromatography
RUO
20532, 29484, 6519
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q07837, Q64319, Q91WV7
SLC3A1
SLC3A1 Fusion Protein Ag9525
150 μL
Primary
Human, Mouse, Rat