missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC3A1, Mouse anti-Human, Clone: 3G3, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00006519-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: FIPNHTSDKHIWFQLSRTRTGKYTDYYIWHDCTHENGKTIPPNNWLSVYGNSSWHFDEVRNQCYFHQFMKEQPDLNFRNPDVQEEIKEILRFWLTKGVDGFSLDAVKFLLSpecifications
| SLC3A1 | |
| Monoclonal | |
| Unconjugated | |
| solute carrier family 3 (cystine, dibasic and neutral amino acid transporters, activator of cystine, dibasic and neutral amino acid transport), member 1 | |
| ATR1/CSNU1/D2H/FLJ34681/NBAT/RBAT | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 3G3 | |
| In 1x PBS, pH 7.4 | |
| BC022386 | |
| SLC3A1 | |
| SLC3A1 (AAH22386, 211 a.a. ∼ 320 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 6519 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction