missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC39A9 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12819222
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12819222 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12819222 Supplier Invitrogen™ Supplier No. PA521074

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is HepG2 cell lysate. PA5-21074 can be used with blocking peptide PEP-1188.

The zinc transporter ZIP9, also known as SLC39A9, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. The zinc transporter family is divided into four subfamilies (I, II, LIV-1 and gufA). ZIP9 is a multipass membrane protein that belongs to the ZIP transporter subfamily I. It is located at the trans-Golgi network regardless of zinc presence and is thought to be a zinc homeostasis regulator acting in the secretory pathway without significantly altering cytosolic zinc homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC39A9
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SLC39A9
Gene Accession No. Q8BFU1, Q9NUM3
Gene Alias 2010002A02Rik; 2610511I23Rik; 4833420E20Rik; AW209151; SLC39A9; solute carrier family 39 (zinc transporter), member 9; solute carrier family 39 member 9; solute carrier family 39, member 9; UNQ714/PRO1377; zinc transporter SLC39A9; zinc transporter ZIP9; ZIP9; ZIP-9; Zrt- and Irt-like protein 9
Gene Symbols SLC39A9
Host Species Rabbit
Immunogen An 18 amino acid peptide near the amino terminus of human ZIP9.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 328133, 55334
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.