missing translation for 'onlineSavingsMsg'
Learn More

SLC39A8/ZIP8 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18414707 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414707 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414707 Supplier Novus Biologicals Supplier No. NBP176505AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC39A8/ZIP8 Polyclonal antibody specifically detects SLC39A8/ZIP8 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC39A8/ZIP8
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias BCG induced integral membrane protein BIGM103, BIGM103BCG-induced integral membrane protein in monocyte clone 103 protein, LIV-1 subfamily of ZIP zinc transporter 6, LZT-Hs6, solute carrier family 39 (metal ion transporter), member 8, solute carrier family 39 (zinc transporter), member 8, Solute carrier family 39 member 8, zinc transporter ZIP8, ZIP8, ZIP-8, Zrt- and Irt-like protein 8
Host Species Rabbit
Immunogen Antibody was raised against a 18 amino acid synthetic peptide near the carboxy terminus of human ZIP8. The immunogen is located within the last 50 amino acids of ZIP8. Amino Acid Squence: PEMDMLREKVTGRKTDF
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 64116
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.