missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC39A8 Polyclonal Antibody

SLC39A8 Polyclonal Antibody
Brand: Invitrogen PA526368
This item is not returnable.
View return policy
Description
SLC39A8 Polyclonal Antibody for Western Blot, Flow
Specifications
| SLC39A8 | |
| Polyclonal | |
| Unconjugated | |
| Slc39a8 | |
| 4933419D20Rik; AA986696; BCG induced integral membrane protein BIGM103; BCG-induced integral membrane protein in monocyte clone 103 protein; BIGM103; CDG2N; LIV-1 subfamily of ZIP zinc transporter 6; LZT-Hs6; PP3105; Slc39a8; solute carrier family 39 (metal ion transporter), member 8; solute carrier family 39 (zinc transporter), member 8; solute carrier family 39 (zinc transporter), member 8 tv2; solute carrier family 39 member 8; Zinc transporter ZIP8; Zip8; ZIP-8; Zrt- and Irt-like protein 8 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 64116 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9C0K1 | |
| Slc39a8 | |
| KLH conjugated synthetic peptide between 218-246 amino acids from the central region of human S39A8. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction