missing translation for 'onlineSavingsMsg'
Learn More

SLC39A7/ZIP7 Antibody, CoraFluor™ 1, Novus Biologicals™

Product Code. 30156713
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30156713 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30156713 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP176504CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC39A7/ZIP7 Polyclonal antibody specifically detects SLC39A7/ZIP7 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunofluorescence,Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC39A7/ZIP7
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Conjugate CoraFluor 1
Formulation PBS
Gene Alias D6S115E, D6S2244EReally interesting new gene 5 protein, H2-KE4, HKE4Solute carrier family 39 member 7, HLA class II region expressed gene KE4, KE4, Ke4 gene, mouse, human homolog of, RING5Histidine-rich membrane protein Ke4, solute carrier family 39 (zinc transporter), member 7, zinc transporter SLC39A7, ZIP7
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 17 amino acids near the amino terminus of human SLC39A7/ZIP7. The immunogen is located within amino acids 20-70 of SLC39A7/ZIP7. Amino Acid Sequence: HDDLQEDFHGHSHRHSH
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7922
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.