missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC39A6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583605
This item is not returnable.
View return policy
Description
Immunogen sequence: HSHHNHAASG KNKRKALCPD HDSDSSGKDP RNSQGKGAHR PEHASGRRNV KDSVSASEVT STVYNTVSEG THFLETIETP RPGKLFPKDV SSSTPPSVTS KSRVSRLAGR KTNESVSEPR KGFMYSRNTN ENPQE Highest antigen sequence indentity to the following orthologs: Rat - 70%, Mouse - 68%.
This gene encodes a protein which interacts with the carboxy-terminus of transcription initiation factor TFIIF, a transcription factor which regulates elongation as well as initiation by RNA polymerase II. The protein may also represent a component of an RNA polymerase II holoenzyme complex. Alternative splicing of this gene results in two transcript variants encoding 2 different isoforms.
Specifications
| SLC39A6 | |
| Polyclonal | |
| Unconjugated | |
| SLC39A6 | |
| EMBL:AAH97493.1}; endoplasmic reticulum membrane protein; endoplasmic reticulum membrane-linked protein; Ermelin; Estrogen-regulated protein LIV-1; LIV1; LIV-1; LIV-1 protein, estrogen regulated; SLC39A6; slc39a6 {ECO:0000312; solute carrier family 39 (metal ion transporter), member 6; solute carrier family 39 (zinc transporter), member 6; solute carrier family 39 member 6; Zinc transporter ZIP6; Zip6; ZIP-6; zrt- and Irt-like protein 6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25800 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13433 | |
| SLC39A6 | |
| Recombinant protein corresponding to Human SLC39A6. Recombinant protein control fragment (Product #RP-91283). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction