missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC39A6 Polyclonal Antibody

Description
Antibody detects endogenous levels of total LIV-1/ZIP6.
This gene encodes a protein which interacts with the carboxy-terminus of transcription initiation factor TFIIF, a transcription factor which regulates elongation as well as initiation by RNA polymerase II. The protein may also represent a component of an RNA polymerase II holoenzyme complex. Alternative splicing of this gene results in two transcript variants encoding 2 different isoforms.
Specifications
Specifications
| Antigen | SLC39A6 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | SLC39A6 |
| Gene Accession No. | Q13433, Q4V887, Q8C145 |
| Gene Alias | EMBL:AAH97493.1}; endoplasmic reticulum membrane protein; endoplasmic reticulum membrane-linked protein; Ermelin; Estrogen-regulated protein LIV-1; LIV1; LIV-1; LIV-1 protein, estrogen regulated; SLC39A6; slc39a6 {ECO:0000312; solute carrier family 39 (metal ion transporter), member 6; solute carrier family 39 (zinc transporter), member 6; solute carrier family 39 member 6; Zinc transporter ZIP6; Zip6; ZIP-6; zrt- and Irt-like protein 6 |
| Gene Symbols | SLC39A6 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?