missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC39A4/ZIP4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82345-0.1ML
This item is not returnable.
View return policy
Description
SLC39A4/ZIP4 Polyclonal specifically detects SLC39A4/ZIP4 in Human samples. It is validated for Western Blot.
Specifications
| SLC39A4/ZIP4 | |
| Polyclonal | |
| PBS, 2% sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 55630 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| acrodermatitis enteropathica, zinc-deficiency type, FLJ20327, MGC74741, solute carrier family 39 (zinc transporter), member 4, Solute carrier family 39 member 4, zinc transporter ZIP4, ZIP-4, ZIP4AEZ, Zrt- and Irt-like protein 4 | |
| The immunogen is a synthetic peptide directed towards the middle region of human SLC39A4 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction