missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SLC39A4/ZIP4 Polyclonal antibody specifically detects SLC39A4/ZIP4 in Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF
Specifications
Specifications
| Antigen | SLC39A4/ZIP4 |
| Applications | Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF |
| Classification | Polyclonal |
| Conjugate | HRP |
| Formulation | PBS |
| Gene Alias | acrodermatitis enteropathica, zinc-deficiency type, FLJ20327, MGC74741, solute carrier family 39 (zinc transporter), member 4, Solute carrier family 39 member 4, zinc transporter ZIP4, ZIP-4, ZIP4AEZ, Zrt- and Irt-like protein 4 |
| Host Species | Goat |
| Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse SLC39A4, Met1-Tyr337, Accession # Q78IQ7 |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?