missing translation for 'onlineSavingsMsg'
Learn More

SLC39A4/ZIP4 Antibody [Allophycocyanin], Novus Biologicals™

Product Code. 30732243 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30732243 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732243 Supplier Novus Biologicals Supplier No. AF7315A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

SLC39A4/ZIP4 Polyclonal antibody specifically detects SLC39A4/ZIP4 in Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC39A4/ZIP4
Applications Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF
Classification Polyclonal
Conjugate APC
Formulation PBS
Gene Alias acrodermatitis enteropathica, zinc-deficiency type, FLJ20327, MGC74741, solute carrier family 39 (zinc transporter), member 4, Solute carrier family 39 member 4, zinc transporter ZIP4, ZIP-4, ZIP4AEZ, Zrt- and Irt-like protein 4
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse SLC39A4, Met1-Tyr337, Accession # Q78IQ7
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 55630
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.