missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC39A3 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76500MFV500
This item is not returnable.
View return policy
Description
SLC39A3 Polyclonal antibody specifically detects SLC39A3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| SLC39A3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| mFluor Violet 500 SE | |
| solute carrier family 39 (zinc transporter), member 3, zinc transporter ZIP3, ZIP-3, ZIP3Solute carrier family 39 member 3, Zrt- and Irt-like protein 3 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the center of human ZIP3. The immunogen is located within amino acids 110 - 160 of ZIP3. Amino Acid Squence: NAGSDVGSDSEYESPF | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 29985 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction