missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC39A2/ZIP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-62490
This item is not returnable.
View return policy
Description
SLC39A2/ZIP2 Polyclonal antibody specifically detects SLC39A2/ZIP2 in Human samples. It is validated for Western Blot
Specifications
| SLC39A2/ZIP2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Eti-1, MGC119190, solute carrier family 39 (zinc transporter), member 2, Solute carrier family 39 member 2, zinc transporter ZIP2, ZIP-2, ZIP2hZIP2, Zrt- and Irt-like protein 2,6A1 | |
| Synthetic peptides corresponding to SLC39A2(solute carrier family 39 (zinc transporter), member 2) The peptide sequence was selected form the N terminal of SLC39A2. Peptide sequence EIESQIQKFMVQNRSASERNSSGDADSAHMEYPYGELIISLGFFFVFFLE. The peptide sequence for this immunogen was taken from within the described region. | |
| 100 μg | |
| Cancer, Cell Cycle and Replication | |
| 29986 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 1.0 μg/mL | |
| Q9NP94 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction