missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC39A14 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£150.00
Specifications
| Antigen | SLC39A14 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL., Immunohistochemistry-Paraffin 2.5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
SLC39A14 Polyclonal specifically detects SLC39A14 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| SLC39A14 | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL., Immunohistochemistry-Paraffin 2.5 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| NP_001128625 | |
| 23516 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the center of human ZIP14. The immunogen is located within amino acids 230 - 280 of ZIP14. Amino Acid Squence: KQKNEHHHGHSHYASE | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| 52 kDa |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| Irt-like protein 14, LIV-1 subfamily of ZIP zinc transporter 4, LZT-Hs4, solute carrier family 39 (metal ion transporter), member 14, solute carrier family 39 (zinc transporter), member 14, solute carrier family 39 member 14, zinc transporter ZIP14, ZIP14, ZIP-14, zrt- and Irt-like protein 14 | |
| SLC39A14 | |
| IgG | |
| Affinity Purified | |
| At least three isoforms of ZIP14 are known to exist; this antibody will detect both isoforms. ZIP14 antibody is predicted to not cross-react with other ZIP family members. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title