missing translation for 'onlineSavingsMsg'
Learn More

SLC39A11 Antibody, DyLight 488, Novus Biologicals™

Product Code. 18433233 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433233 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18433233 Supplier Novus Biologicals Supplier No. NBP177344G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC39A11 Polyclonal antibody specifically detects SLC39A11 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC39A11
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias C17orf26, chromosome 17 open reading frame 26, solute carrier family 39 (metal ion transporter), member 11, Solute carrier family 39 member 11, zinc transporter ZIP11, ZIP11, ZIP-11, Zrt- and Irt-like protein 11
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide near the center of the human ZIP11. The immunogen is located within amino acids 130 - 180 of ZIP11. Amino Acid Squence: IDKDENGEAYQRKKA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 201266
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.