missing translation for 'onlineSavingsMsg'
Learn More

SLC39A10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£222.00 - £470.00

Specifications

Antigen SLC39A10
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18610796
View Documents
Novus Biologicals
NBP2-38306-25ul
25 μL
£222.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18148737
View Documents
Novus Biologicals
NBP2-38306
0.1 mL
£470.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SLC39A10 Polyclonal specifically detects SLC39A10 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

SLC39A10
Polyclonal
Rabbit
Human
Q9ULF5
57181
This antibody was developed against a recombinant protein corresponding to amino acids: KHYKQQRGKQKWFMKQNTEESTIGRKLSDHKLNNTPDSDWLQLKPLAGTDDSVVSEDRLNETELTDLEGQQESPPKNYLCIEEEKIIDHSHSDGL
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
DKFZp564L2123, DKFZp781L10106, FLJ90515, KIAA1265, LZT-Hs2, MGC126565, MGC138428, solute carrier family 39 (metal ion transporter), member 10, solute carrier family 39 (zinc transporter), member 10, Solute carrier family 39 member 10, zinc transporter ZIP10, ZIP10, ZIP-10, Zrt- and Irt-like protein 10
SLC39A10
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.