missing translation for 'onlineSavingsMsg'
Learn More

SLC39A10 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18431287 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431287 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431287 Supplier Novus Biologicals Supplier No. NBP176507AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC39A10 Polyclonal antibody specifically detects SLC39A10 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC39A10
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias DKFZp564L2123, DKFZp781L10106, FLJ90515, KIAA1265, LZT-Hs2, MGC126565, MGC138428, solute carrier family 39 (metal ion transporter), member 10, solute carrier family 39 (zinc transporter), member 10, Solute carrier family 39 member 10, zinc transporter ZIP10, ZIP10, ZIP-10, Zrt- and Irt-like protein 10
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide near the center of human ZIP10. The immunogen is located within amino acids 550 - 600 of ZIP10. Amino Acid Squence: AGTDDSVVSEDRLNETEL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 57181
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.