missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC38A5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561506
This item is not returnable.
View return policy
Description
Immunogen sequence: MELQDPKMNG ALPSDAVGYR QEREGFLPSR GPAPGSKPVQ FMDFE Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 72%.
The protein encoded by this gene is a system N sodium-coupled amino acid transporter involved in the transfer of glutamine, asparagine, histidine, serine, alanine, and glycine. The encoded protein does not transport charged amino acids.
Specifications
| SLC38A5 | |
| Polyclonal | |
| Unconjugated | |
| SLC38A5 | |
| amino acid transport system N2; amino acid transporter system N2; C81234; E330031E14; JM24; PP7194; SLC38A5; Sn2; Snat5; sodium-coupled neutral amino acid transporter 5; sodium-coupled neutral amino acid transporter 5; LOW QUALITY PROTEIN: sodium-coupled neutral amino acid transporter 5; solute carrier family 38 (amino acid transporter), member 5; solute carrier family 38 member 5; solute carrier family 38, member 5; system N transporter 2; transport system N, protein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 92745 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WUX1 | |
| SLC38A5 | |
| Recombinant protein corresponding to Human SLC38A5. Recombinant protein control fragment (Product #RP-98325). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction