missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC38A4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20857-1-AP-150UL
This item is not returnable.
View return policy
Description
SLC38A4 is found predomitly in liver and transports both cationic and neutral amino acids. The transport of cationic amino acids by SLC38A4 is Na(+) and pH independent, while the transport of neutral amino acids is Na(+) and pH dependent.The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. Four alternatively spliced transcript variants encoding the same protein have been found for this gene.Specifications
| SLC38A4 | |
| Polyclonal | |
| Unconjugated | |
| SLC38A4 | |
| Amino acid transporter A3, ATA3, NAT3, PAAT, SLC38A4, SNAT4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55089 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q969I6 | |
| SLC38A4 | |
| SLC38A4 Fusion Protein Ag14827 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction