missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC38A4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572879
This item is not returnable.
View return policy
Description
SLC38A4 Polyclonal Antibody for Western Blot, IHC (P), ELISA
SLC38A4 is found predominantly in liver and transports both cationic and neutral amino acids. The transport of cationic amino acids by SLC38A4 is Na(+) and pH independent, while the transport of neutral amino acids is Na(+) and pH dependent.The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. Four alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
| SLC38A4 | |
| Polyclonal | |
| Unconjugated | |
| SLC38A4 | |
| 1110012E16Rik; 1700012A18Rik; amino acid transport system A3; amino acid transporter A3; amino acid transporter system A3; Ata3; FLJ10191; mATA3; MGC126876; mNAT3; N amino acid transporter 3; Na(+)-coupled neutral amino acid transporter 4; NAT3; PAAT; Slc38a4; Snat4; sodium-coupled neutral amino acid transporter 4; Solute carrier family 38 member 4; solute carrier family 38, member 4; system A amino acid transporter 3; System N amino acid transporter 3; system-N amino acid transporter 3; Unknown (protein for MGC:113830); wu:fd51c05; zgc:103694; zgc:113830 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55089, 69354 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q8R1S9, Q969I6 | |
| SLC38A4 | |
| SLC38A4 antibody for an 18 amino acid peptide from near the center of human SLC38A4. The immunogen is within amino acids 270 - 320 of SLC38A4. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction